Order Entry
Export
ContactUsLinkComponent
Anti-Fibroblast Growth Factor Receptor 1 beta Mouse monoclonal antibody
Anti-Fibroblast Growth Factor Receptor 1 beta Mouse monoclonal antibody
Catalog # QEDB30106
Supplier:  QED Bioscience
Anti-Fibroblast Growth Factor Receptor 1 beta Mouse monoclonal antibody
Catalog # QEDB30106
Supplier:  QED Bioscience

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast growth factor receptor 1B
  • Antigen symbol:
    FGFR1B
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pk:
    250 µG

Specifications

About this item

Type: Primary
Antigen: Fibroblast Growth Factor Receptor 1 beta
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human