Order Entry
Export
ContactUsLinkComponent
 
Anti-FBXO17 Rabbit Polyclonal Antibody
  QEDB28105
Anti-FBXO17 Rabbit Polyclonal Antibody
  QEDB28105

 

  • Antibody type:
    Primary
  • Antigen name:
    F-box protein 17
  • Antigen symbol:
    FBXO17
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: FBXO17 (F-box protein 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human