Anti-Streptococcus pneumoniae Mouse Monoclonal Antibody
Catalog # QEDB22004
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen symbol:Streptococcus pneumoniae
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgM
- Cross adsorption:none
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: Streptococcus pneumoniae
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: