Anti-High MW Kininogen Light Chain Mouse Monoclonal Antibody
Catalog # QEDB20001
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:High MW Kininogen Light Chain
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: High MW Kininogen Light Chain
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human