Order Entry
Export
ContactUsLinkComponent
Anti-F5 Mouse monoclonal antibody unconjugated
Anti-F5 Mouse monoclonal antibody unconjugated
  QEDB19702
Anti-F5 Mouse monoclonal antibody unconjugated
  QEDB19702

 

  • Antibody type:
    Primary
  • Antigen name:
    coagulation factor V (proaccelerin, labile factor)
  • Antigen symbol:
    F5
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pk:
    250 µG

 

 

Type: Primary
Antigen: F5 (coagulation factor V (proaccelerin, labile factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human