Order Entry
Export
ContactUsLinkComponent
 
Anti-Mycobacterium avium Mouse Monoclonal Antibody
  QEDB18103
Anti-Mycobacterium avium Mouse Monoclonal Antibody
  QEDB18103

 

  • Antibody type:
    Primary
  • Antigen name:
    Mycobacterium avium
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 mg
  • Pk:
    500 µG

 

 

Type: Primary
Antigen: Mycobacterium avium
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: