Order Entry
Export
ContactUsLinkComponent
Anti-Rabbit Immunoglobulin Mouse Monoclonal Antibody
Anti-Rabbit Immunoglobulin Mouse Monoclonal Antibody
Catalog # QEDB14602
Supplier:  QED Bioscience
Anti-Rabbit Immunoglobulin Mouse Monoclonal Antibody
Catalog # QEDB14602
Supplier:  QED Bioscience

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Rabbit Immunoglobulin
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Rabbit
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 mg
  • Pk:
    500 µG

Specifications

About this item

Type: Primary
Antigen: Rabbit Immunoglobulin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rabbit