Anti-Glucose Oxidase Mouse Monoclonal Antibody
QEDB13101
- Antibody type:Primary
- Antigen name:Glucose Oxidase
- Antigen symbol:GOX
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:500 µG
Type: Primary
Antigen: Glucose Oxidase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: