Anti-Nav1.8 Na+ Channel Mouse Monoclonal Antibody
Catalog # QEDB11523
Supplier: QED Bioscience
Specifications
- Antibody type:Primary
- Antigen name:Nav1.8 Na+ Channel
- Antigen symbol:NAV1.8
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Nav1.8 Na+ Channel
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat