Order Entry
Export
ContactUsLinkComponent
 
Anti-HSPA13 Mouse Monoclonal Antibody [clone: 3A3]
  QEDB11117
Anti-HSPA13 Mouse Monoclonal Antibody [clone: 3A3]
  QEDB11117

 

  • Antibody type:
    Primary
  • Antigen name:
    Heat shock protein 70kDa family, member 13
  • Antigen symbol:
    HSPA13
  • Clonality:
    Monoclonal
  • Clone:
    3A3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: monoclonal
Clone: 3A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: