Anti-HSPA13 Mouse Monoclonal Antibody [clone: 2A4]
QEDB11116
- Antibody type:Primary
- Antigen name:Heat shock protein 70kDa family, member 13
- Antigen symbol:HSPA13
- Clonality:Monoclonal
- Clone:2A4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: monoclonal
Clone: 2A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: