Order Entry
Export
ContactUsLinkComponent
Anti-CRYAB Rabbit Polyclonal Antibody
Anti-CRYAB Rabbit Polyclonal Antibody
Catalog # QEDB11073
Supplier:  QED Bioscience
Anti-CRYAB Rabbit Polyclonal Antibody
Catalog # QEDB11073
Supplier:  QED Bioscience

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Crystallin, alpha B
  • Antigen symbol:
    CRYAB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: