Order Entry
Export
ContactUsLinkComponent
Anti-GST Mouse Monoclonal Antibody (Biotin)
Anti-GST Mouse Monoclonal Antibody (Biotin)
Catalog # GENSA00867-200
Supplier:  Genscript
undefined
Anti-GST Mouse Monoclonal Antibody (Biotin)
Catalog # GENSA00867-200
Supplier:  Genscript

Specifications

  • Antibody type:
    Tag
  • Antigen symbol:
    GST
  • Clonality:
    Monoclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    5x40 µg
  • Pk:
    200 µG

Specifications

About this item

Type: Tag
Antigen: GST
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: