Anti-ACTB Mouse Monoclonal Antibody
Supplier: Genscript
Certificates
About this item
Type: Tag
Antigen: ACTB (actin, beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Specifications
- Catalog No:
- GENSA00702-100
- GENSA00702-200
- Antigen symbol:
- ACTB
- ACTB
- Antigen name:
- Actin beta
- Actin beta
- Antigen synonyms:
- PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
- PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
- Antibody type:
- Tag
- Tag
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice