Anti-CAPN1 Goat Polyclonal Antibody
EVBIEB11988
- Antibody type:Primary
- Antigen name:calpain 1, (mu/I) large subunit
- Antigen symbol:CAPN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: CAPN1 (calpain 1, (mu/I) large subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human