Anti-Mucin 19/ Apomucin Goat Polyclonal Antibody
EVBIEB10619
- Antibody type:Primary
- Antigen name:Mucin 19
- Antigen symbol:MUC19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Epitope:C-terminal
- Cross adsorption:none
- Antigen synonyms:Apomucin
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: Mucin 19 / Apomucin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat