Anti-BOK Goat polyclonal antibody unconjugated
Catalog # EVBIEB10361
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:BCL2-related ovarian killer
- Antigen symbol:BOK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: BOK (BCL2-related ovarian killer)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: