Order Entry
Export
ContactUsLinkComponent
Anti-CIRBP Goat polyclonal antibody unconjugated
Anti-CIRBP Goat polyclonal antibody unconjugated
  EVBIEB10235
undefined
Anti-CIRBP Goat polyclonal antibody unconjugated
  EVBIEB10235

 

  • Antibody type:
    Primary
  • Antigen name:
    cold inducible RNA binding Protein
  • Antigen symbol:
    CIRBP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Amino acid number:
    81 to 91
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: CIRBP (cold inducible RNA binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: