Anti-PIK3C2A Goat Polyclonal Antibody
EVBIEB06291
- Antibody type:Primary
- Antigen name:phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 alpha
- Antigen symbol:PIK3C2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Mouse
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: PIK3C2A (phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse