Anti-NFATC4 Goat Polyclonal Antibody
Catalog # EVBIEB05562
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4
- Antigen symbol:NFATC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Dog,Rat,Mouse
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: NFATC4 (nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Pig, Rat