Anti-TLR10 Rabbit Polyclonal Antibody
BSBTPA1958
- Antibody type:Primary
- Antigen name:Toll-like Receptor 10
- Antigen symbol:TLR10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Toll-like receptor 10|CD290
- Amino acid number:88 to 102
- Sequence:DLKTFEFNKELRYLD
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human TLR10 (88-102aa DLKTFEFNKELRYLD).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Toll-like receptor 10(TLR10) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: TLR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human