Anti-GCSF Receptor Rabbit Polyclonal Antibody
BSBTPA1906
- Antibody type:Primary
- Antigen name:Colony Stimulating Factor 3 Receptor (granulocyte)
- Antigen symbol:GCSFR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:254 to 271
- Sequence:EPWQPGLHINQKCELRHK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human GCSF Receptor (254-271aa EPWQPGLHINQKCELRHK).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Granulocyte colony-stimulating factor receptor(CSF3R) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: GCSF Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human