Anti-CD147 Rabbit Polyclonal Antibody
Catalog # BSBTPA1526
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Basigin
- Antigen symbol:CD147
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CD147|collagenase stimulatory factor|basigin|TCSF|tumor cell-derived collagenase stimulatory factor|5F7|leukocyte activation antigen M6|OK blood group antigen|EMMPRIN|OK|CD147 antigen|extracellular matrix metalloproteinase inducer|M6
- Amino acid number:186 to 203
- Sequence:QKTEFKVDSDDQWGEYSC
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human CD147 (186-203aa QKTEFKVDSDDQWGEYSC).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Basigin(BSG) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: CD147
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human