Anti-XIAP Rabbit Polyclonal Antibody
Catalog # BSBTPA1512
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:X-linked inhibitor of apoptosis
- Antigen symbol:XIAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:IAP-like protein|HILP|API3|Baculoviral IAP repeat-containing protein 4|X-linked IAP|X-linked inhibitor of apoptosis protein|XIAP|IAP3|Inhibitor of apoptosis protein 3
- Amino acid number:14 to 34
- Sequence:ADINKEEEFVEEFNRLKTFAN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human XIAP (14-34aa ADINKEEEFVEEFNRLKTFAN), different from the related mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for E3 ubiquitin-protein ligase XIAP(XIAP) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: XIAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat