Order Entry
Export
ContactUsLinkComponent
 
Anti-PSTAIR Mouse Monoclonal Antibody [clone: IL-16]
Catalog # BSBTMA1087
Supplier:  Boster Bio
Anti-PSTAIR Mouse Monoclonal Antibody [clone: IL-16]
Catalog # BSBTMA1087
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PSTAIR
  • Clonality:
    Monoclonal
  • Clone:
    IL-16
  • Conjugation:
    BSA (Bovine Serum Albumin)
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic 16 amino acid oligopeptide containing the PSTAIR sequence conjugated to BSA.
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: PSTAIR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse