Anti-NCAM Mouse Monoclonal Antibody [clone: IML-43]
Catalog # BSBTMA1068
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Neural Cell Adhesion Molecule 1
- Antigen symbol:NCAM1
- Clonality:Monoclonal
- Clone:IML-43
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:NCAM|MSK39|CD56
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Growth cone enriched plasma membrane fraction from E17 rat forebrain.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: NCAM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat