Anti-P-Glycoprotein (MDR) Mouse Monoclonal Antibody [clone: PG-13]
Catalog # BSBTMA1060
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:P-Glycoprotein (MDR)
- Clonality:Monoclonal
- Clone:PG-13
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Mixture of human and hamster drug-resistant whole cells and crude plasma membranes.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: P-Glycoprotein (MDR)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human