Order Entry
Export
ContactUsLinkComponent
Anti-Catenin gamma (Plakoglobin) Mouse Monoclonal Antibody [clone: CN-3]
Anti-Catenin gamma (Plakoglobin) Mouse Monoclonal Antibody [clone: CN-3]
Catalog # BSBTMA1012
Supplier:  Boster Bio
undefined
Anti-Catenin gamma (Plakoglobin) Mouse Monoclonal Antibody [clone: CN-3]
Catalog # BSBTMA1012
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cathepsin G
  • Antigen symbol:
    CATG
  • Clonality:
    Monoclonal
  • Clone:
    CN-3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Bovine
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Antigen synonyms:
    CATG|CG
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant chicken plakoglobin.
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Mouse IgG monoclonal antibody for Catenin gamma (Plakoglobin), junction plakoglobin ( JUP) detection. Tested with WB, IHC-F, ICC in Human; chicken; rat. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: Catenin gamma (Plakoglobin)
Clonality: Monoclonal
Clone: CN-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine