Order Entry
Export
ContactUsLinkComponent
Anti-FPR2 Rabbit Polyclonal Antibody (DyLight® 650)
Anti-FPR2 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNLS1878C
Supplier:  Novus Biologicals
Anti-FPR2 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNLS1878C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Formyl Peptide Receptor 2
  • Antigen symbol:
    FPR2
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 650
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    2358
  • Antigen synonyms:
    FPRH1|FPRH2|FMLPX|FPRL1|FPR2A|FMLP-R-II|HM63|LXA4R|ALXR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µL
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: FPR2 (formyl peptide receptor 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat