Order Entry
Export
ContactUsLinkComponent
Anti-MAS1 Rabbit Polyclonal Antibody
Anti-MAS1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-MAS1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNLS1531
  • NOVUNLS1531SS
  • Antigen symbol:
  • MAS1
  • MAS1
  • Antigen name:
  • MAS1 oncogene
  • MAS1 oncogene
  • Antigen synonyms:
  • MGC119966|MGRA|MAS
  • MGC119966|MGRA|MAS
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 4142
  • 4142
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information