Anti-HMGB1 Mouse Monoclonal Antibody [clone: 19N15F4]
Catalog # NOVUNBP2-27401
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:high mobility group box 1
- Antigen symbol:HMGB1
- Clonality:Monoclonal
- Clone:19N15F4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:3146
- Antigen synonyms:SBP-1|HMG3|HMG1|HMG-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: HMGB1 (high mobility group box 1)
Clonality: monoclonal
Clone: 19N15F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse