Anti-BAG3 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27398
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:BCL2-associated athanogene 3
- Antigen symbol:BAG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:9531
- Antigen synonyms:MFM6|CAIR-1|BAG-3|BIS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: BAG3 (BCL2-associated athanogene 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse