Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 32A1380]
NOVUNBP2-27350
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 10a
- Antigen symbol:TNFRSF10A
- Clonality:Monoclonal
- Clone:32A1380
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Gene ID:8797
- Antigen synonyms:TRAILR1|DR4|APO2|CD261|TRAILR-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: TNFRSF10A (tumor necrosis factor receptor superfamily, member 10a)
Clonality: monoclonal
Clone: 32A1380
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human