Anti-OLIG2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27301
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:oligodendrocyte lineage transcription factor 2
- Antigen symbol:OLIG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Gene ID:10215
- Antigen synonyms:RACK17|bHLHe19|PRKCBP2|BHLHB1|OLIGO2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: OLIG2 (oligodendrocyte lineage transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human