Anti-NEUROG3 Rabbit Polyclonal Antibody
NOVUNBP2-27115
- Antibody type:Primary
- Antigen name:Neurogenin 3
- Antigen symbol:NEUROG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Gene ID:50674
- Antigen synonyms:bHLHa7|Atoh5|NGN-3|ngn3|Math4B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: NEUROG3 (neurogenin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human