Anti-MAF Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24551
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog
- Antigen symbol:MAF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Gene ID:4094
- Antigen synonyms:c-MAF|AYGRP|CCA4|CTRCT21
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: MAF (v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat