Anti-TLR4 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24538
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Toll-like Receptor 4
- Antigen symbol:TLR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:7099
- Antigen synonyms:ARMD10, CD284, TLR-4, TOLL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: