Anti-TRPV1 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-97417R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Transient receptor potential cation channel subfamily V member 1
- Antigen symbol:TRPV1
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Gene ID:7442
- Antigen synonyms:TrpV1|VR1|TRPV1|Transient receptor potential cation channel subfamily V member 1|Capsaicin receptor
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: TRPV1 (transient receptor potential cation channel, subfamily V, member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: