Anti-BTK Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The BTK [p Tyr223] Antibody from Novus Biologicals is a rabbit polyclonal antibody to BTK. This antibody reacts with human, mouse. The BTK [p Tyr223] Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: BTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-78295SS
- NOVUNBP1-78295
- Antigen symbol:
- BTK
- BTK
- Antigen name:
- Bruton agammaglobulinemia tyrosine kinase
- Bruton agammaglobulinemia tyrosine kinase
- Antigen synonyms:
- XLA|AGMX1|RP1-164F3.2|ATK|PSCTK1|MGC126262|BPK|MGC126261|AT|IMD1
- XLA|AGMX1|RP1-164F3.2|ATK|PSCTK1|MGC126262|BPK|MGC126261|AT|IMD1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 695
- 695
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice