Anti-Lamin A + C R453W Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12A-2F5]
Catalog # NOVUNBP1-77401PE
Supplier: Novus Biologicals
Specifications
- Antigen synonyms:LMNL1|FPLD|FPL|IDC|PRO1|LMNC|CMT2B1|LGMD1B|LMN1|CDCD1|LFP|FPLD2|LDP1|CDDC|CMD1A|EMD2|HGPS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Cat. no.:NOVUNBP1-77401PE
- Pk:100 Tests