Anti-HSPD1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Rabbit Polyclonal Hsp60 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Chicken, Hamster, Primate.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: HSP60
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- NOVUNBP1-77397SS
- NOVUNBP1-77397
- Antigen symbol:
- HSP60
- HSP60
- Antigen name:
- Heat Shock Protein 60
- Heat Shock Protein 60
- Antigen synonyms:
- Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
- Mytonin Chaperone 60|GROEL|HSP65|P60 lymphocyte protein|HLD4|HSP-60|HuCHA60|mitochondrial matrix protein P1|chaperonin 60|HSP60|mitochondrial|CPN60|short heat shock protein 60 Hsp60s1|heat shock protein 65|60 kDa chaperonin|60 kDa heat shock protein|SPG13
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 3329
- 3329
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice