Order Entry
Export
ContactUsLinkComponent
 
Anti-HSPD1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-77396G
Supplier:  Novus Biologicals
Anti-HSPD1 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-77396G
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Heat shock 60kDa protein 1
  • Antigen symbol:
    HSPD1
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Gene ID:
    3329
  • Antigen synonyms:
    GROEL|HSP65|HLD4|HSP-60|HuCHA60|Chaperonin 60|HSP60|Mitochondrial Matrix Protein P1|CPN60|Heat Shock Protein 65|SPG13
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: HSPD1 (heat shock 60kDa protein 1 (chaperonin))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: