Anti-CHD7 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNBP1-77393V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Chromodomain helicase DNA binding protein 7
- Antigen symbol:CHD7
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:55636
- Antigen synonyms:CRG|HH5|KAL5|IS3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CHD7 (chromodomain helicase DNA binding protein 7)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse