Anti-CHD7 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-77393R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Chromodomain helicase DNA binding protein 7
- Antigen symbol:CHD7
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:55636
- Antigen synonyms:HH5|KAL5|ATP-dependent helicase CHD7|chromodomain helicase DNA binding protein 7 isoform CRA_e|IS3|chromodomain-helicase-DNA-binding protein 7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CHD7 (chromodomain helicase DNA binding protein 7)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse