Anti-ATF6 Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNBP1-75478C
- Antibody type:Primary
- Antigen name:Activating transcription factor 6
- Antigen symbol:ATF6
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Gene ID:22926
- Antigen synonyms:ATF6A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: ATF6 (activating transcription factor 6)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human