Order Entry
Export
ContactUsLinkComponent
Anti-POU5F1 Mouse Monoclonal Antibody [clone: 7E7]
Anti-POU5F1 Mouse Monoclonal Antibody [clone: 7E7]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-POU5F1 Mouse Monoclonal Antibody [clone: 7E7]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-51664SS
  • NOVUNBP1-51664
  • Antigen symbol:
  • POU5F1
  • POU5F1
  • Antigen name:
  • POU class 5 homeobox 1
  • POU Class 5 Homeobox 1
  • Antigen synonyms:
  • OCT4|Oct-4|OCT3|OTF3|OTF-3|OTF4|Oct-3
  • POU domain class 5 transcription factor 1|Oct-4|OCT3|OTF3.|Octamer-binding transcription factor 3|Oct-3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 7E7
  • 7E7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5460
  • 5460
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information