Order Entry
Export
ContactUsLinkComponent
Anti-GOLM1 Rabbit Polyclonal Antibody
Anti-GOLM1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-GOLM1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-50627SS
  • NOVUNBP1-50627
  • Antigen symbol:
  • GOLM1
  • GOLM1
  • Antigen name:
  • Golgi membrane protein 1
  • Golgi membrane protein 1
  • Antigen synonyms:
  • GP73|FLJ22634|golgi membrane protein GP73|golgi membrane protein 1|C9orf155|GOLPH2|GOLM1|FLJ23608|golgi phosphoprotein 2|bA379P1.3|golgi protein|PSEC0257|RP11-379P1.3|73-kD
  • GP73|FLJ22634|golgi membrane protein GP73|golgi membrane protein 1|C9orf155|GOLPH2|GOLM1|FLJ23608|golgi phosphoprotein 2|bA379P1.3|golgi protein|PSEC0257|RP11-379P1.3|73-kD
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 51280
  • 51280
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information