Anti-DUX4 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: P4H2]
Catalog # NOVUNBP1-49552F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Double homeobox 4 like 1
- Antigen symbol:DUX4L1
- Clonality:Monoclonal
- Clone:P4H2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:22947
- Antigen synonyms:DUX10|DUX4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: DUX4 (double homeobox 4)
Clonality: monoclonal
Clone: P4H2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human