Order Entry
Export
ContactUsLinkComponent
Anti-DUX4 Mouse Monoclonal Antibody [clone: P4H2]
Anti-DUX4 Mouse Monoclonal Antibody [clone: P4H2]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-DUX4 Mouse Monoclonal Antibody [clone: P4H2]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-49552
  • NOVUNBP1-49552SS
  • Antigen symbol:
  • DUX4
  • DUX4
  • Antigen name:
  • Double homeobox 4
  • Double homeobox 4
  • Antigen synonyms:
  • double homeobox protein 10|DUX10|double homeobox protein 4/10|double homeobox protein DUX10|double homeobox protein 4
  • double homeobox protein 10|DUX10|double homeobox protein 4/10|double homeobox protein DUX10|double homeobox protein 4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • P4H2
  • P4H2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 22947
  • 22947
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information