Anti-RNA polymerase II Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNBP1-49546V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (RNA) II (DNA directed) polypeptide A, 220kDa
- Antigen symbol:POLR2A
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- Western blot:Yes
- Gene ID:5430
- Antigen synonyms:hsRPB1|RPOL2|POLRA|RPBh1|RpIILS|RPO2|RPB1|POLR2|hRPB220
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: RNA polymerase II
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: